Product Details

SNP ID
rs11556497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41264516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCCCGGGCCATGGATGTGCGC[C/T]GTCTGAAGGTGAACGAACTTCGCGA
Phenotype
MIM: 109135 MIM: 605800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AXL PubMed Links

Gene Details

Gene
AXL
Gene Name
AXL receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
HNRNPUL1
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301016.2 343 Intron NP_001287945.1
NM_001321208.1 343 UTR 5 NP_001308137.1
NM_001321211.1 343 Intron NP_001308140.1
NM_007040.5 343 Missense Mutation CGT,TGT R5C NP_008971.2
NM_144732.4 343 Intron NP_653333.1
XM_005258459.3 343 Missense Mutation CGT,TGT R5C XP_005258516.1
XM_005258461.4 343 Intron XP_005258518.1
XM_005258463.3 343 UTR 5 XP_005258520.1
XM_005258464.3 343 Intron XP_005258521.1
XM_005258465.4 343 Intron XP_005258522.1
XM_011526392.2 343 Missense Mutation CGT,TGT R5C XP_011524694.1
XM_011526393.2 343 Intron XP_011524695.1
XM_011526394.2 343 Intron XP_011524696.1
XM_011526395.2 343 Intron XP_011524697.1

View Full Product Details