Product Details

SNP ID
rs8104417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52976891 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGATACTAGTATGTCTTCTCTGG[G/T]GTCTCATGTGTGCCAGGGTCCTATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF702P PubMed Links

Gene Details

Gene
ZNF702P
Gene Name
zinc finger protein 702, pseudogene
There are no transcripts associated with this gene.

View Full Product Details