Product Details

SNP ID
rs7260332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1880018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCGTAGCCGGAGTAGTCGTAGG[A/T]GAAGATGTTGCAGTGGAGGCGGGAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABHD17A PubMed Links

Gene Details

Gene
ABHD17A
Gene Name
abhydrolase domain containing 17A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130111.1 3708 Intron NP_001123583.1
NM_031213.3 3708 Intron NP_112490.3
XM_011528338.1 3708 Missense Mutation ACC,TCC T195S XP_011526640.1
XM_017027341.1 3708 Missense Mutation ACC,TCC T144S XP_016882830.1
XM_017027342.1 3708 UTR 5 XP_016882831.1

View Full Product Details