Product Details
- SNP ID
-
rs11556474
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63873779 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCTGGGGGAGCTGAAACAGAAC[C/G]TGTCCAGGAGCTGGCATGACGTGCA
- Phenotype
-
MIM: 603747
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TPD52L2
PubMed Links
Gene Details
- Gene
- TPD52L2
- Gene Name
- tumor protein D52 like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001243891.1 |
415 |
Missense Mutation |
CTG,GTG |
L68V |
NP_001230820.1 |
NM_001243892.1 |
415 |
Missense Mutation |
CTG,GTG |
L70V |
NP_001230821.1 |
NM_001243894.1 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_001230823.1 |
NM_001243895.1 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_001230824.1 |
NM_003288.3 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_003279.2 |
NM_199359.2 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_955391.1 |
NM_199360.2 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_955392.1 |
NM_199361.2 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_955393.1 |
NM_199362.2 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_955394.1 |
NM_199363.2 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
NP_955395.1 |
XM_011529034.2 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
XP_011527336.1 |
XM_011529035.2 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
XP_011527337.1 |
XM_017028038.1 |
415 |
Missense Mutation |
CTG,GTG |
L93V |
XP_016883527.1 |
View Full Product Details