Product Details
- SNP ID
-
rs9624836
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:25450120 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGATATGTATTTTTTATGCTATTTG[G/T]TTTTTTTTTCTGTGTATATGTTTGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CRYBB2P1
PubMed Links
Gene Details
- Gene
- CRYBB2P1
- Gene Name
- crystallin beta B2 pseudogene 1
There are no transcripts associated with this gene.
- Gene
- MIR6817
- Gene Name
- microRNA 6817
There are no transcripts associated with this gene.
View Full Product Details