Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001024666.2 | 2746 | UTR 3 | NP_001019837.1 | ||
NM_001184960.1 | 2746 | UTR 3 | NP_001171889.1 | ||
NM_031892.2 | 2746 | UTR 3 | NP_114098.1 | ||
XM_005274494.2 | 2746 | UTR 3 | XP_005274551.1 | ||
XM_011545498.2 | 2746 | UTR 3 | XP_011543800.1 | ||
XM_011545499.2 | 2746 | UTR 3 | XP_011543801.1 | ||
XM_011545500.2 | 2746 | UTR 3 | XP_011543802.1 | ||
XM_011545502.2 | 2746 | UTR 3 | XP_011543804.1 | ||
XM_011545503.2 | 2746 | Intron | XP_011543805.1 | ||
XM_017029459.1 | 2746 | UTR 3 | XP_016884948.1 | ||
XM_017029460.1 | 2746 | UTR 3 | XP_016884949.1 | ||
XM_017029461.1 | 2746 | UTR 3 | XP_016884950.1 | ||
XM_017029462.1 | 2746 | UTR 3 | XP_016884951.1 | ||
XM_017029463.1 | 2746 | UTR 3 | XP_016884952.1 | ||
XM_017029464.1 | 2746 | UTR 3 | XP_016884953.1 | ||
XM_017029465.1 | 2746 | UTR 3 | XP_016884954.1 | ||
XM_017029466.1 | 2746 | UTR 3 | XP_016884955.1 | ||
XM_017029467.1 | 2746 | UTR 3 | XP_016884956.1 | ||
XM_017029468.1 | 2746 | UTR 3 | XP_016884957.1 | ||
XM_017029469.1 | 2746 | UTR 3 | XP_016884958.1 | ||
XM_017029470.1 | 2746 | UTR 3 | XP_016884959.1 |