Product Details

SNP ID
rs10284116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:20140695 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAATAATCAAAGGAGACTACTTA[C/T]GGACTATGAAGTATGCTCCTCTCCT
Phenotype
MIM: 300186 MIM: 300075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF1AX PubMed Links

Gene Details

Gene
EIF1AX
Gene Name
eukaryotic translation initiation factor 1A, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001412.3 Intron NP_001403.1
Gene
EIF1AX-AS1
Gene Name
EIF1AX antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPS6KA3
Gene Name
ribosomal protein S6 kinase A3
There are no transcripts associated with this gene.

Gene
SCARNA9L
Gene Name
small Cajal body-specific RNA 9-like
There are no transcripts associated with this gene.

View Full Product Details