Product Details

SNP ID
rs11538308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:2854666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAATCCCTCACCTGGTGACTCAT[C/G]ATGCTCGAACCATCCGCTACCCAGA
Phenotype
MIM: 470000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPS4Y1 PubMed Links

Gene Details

Gene
RPS4Y1
Gene Name
ribosomal protein S4, Y-linked 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008.3 Intron NP_000999.1
XM_017030061.1 Intron XP_016885550.1

View Full Product Details