Product Details

SNP ID
rs10017282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87426216 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTTCCAGAAGGATCTACTGGTGA[C/T]GTGTGTAGAATATTGTTTTATTCCT
Phenotype
MIM: 606022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT9 PubMed Links

Gene Details

Gene
NUDT9
Gene Name
nudix hydrolase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248011.1 Intron NP_001234940.1
NM_024047.4 Intron NP_076952.1
NM_198038.2 Intron NP_932155.1
XM_011532031.2 Intron XP_011530333.1
XM_011532032.2 Intron XP_011530334.1

View Full Product Details