Product Details

SNP ID
rs7441958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112543092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAATTGTGAACTAGTTCAAATA[C/T]ACAGAAAGCAAAGAAACAATATTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZGRF1 PubMed Links

Gene Details

Gene
ZGRF1
Gene Name
zinc finger GRF-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018392.4 Intron NP_060862.3
XM_005263115.3 Intron XP_005263172.1
XM_011532091.2 Intron XP_011530393.1
XM_011532092.2 Intron XP_011530394.1
XM_011532093.2 Intron XP_011530395.1
XM_011532094.2 Intron XP_011530396.1
XM_011532096.2 Intron XP_011530398.1
XM_011532097.2 Intron XP_011530399.1
XM_011532098.2 Intron XP_011530400.1
XM_011532099.2 Intron XP_011530401.1
XM_017008369.1 Intron XP_016863858.1
XM_017008370.1 Intron XP_016863859.1
XM_017008371.1 Intron XP_016863860.1
XM_017008372.1 Intron XP_016863861.1
XM_017008373.1 Intron XP_016863862.1

View Full Product Details