Product Details

SNP ID
rs11541660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:137521111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCATGAGTTCGTGTTTCCCATCA[A/T]TGAGGTGGTTGAGCACATTGTCAAA
Phenotype
MIM: 608287
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDH18 PubMed Links

Gene Details

Gene
PCDH18
Gene Name
protocadherin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300828.1 3736 Missense Mutation AAT,ATT N1108I NP_001287757.1
NM_019035.4 3736 Missense Mutation AAT,ATT N1109I NP_061908.1
XM_006714239.3 3736 Intron XP_006714302.1
XM_017008311.1 3736 UTR 3 XP_016863800.1

View Full Product Details