Product Details

SNP ID
rs6850272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:157219144 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTAGGACTACATCAAAAAGAGG[G/T]AAAGGTTATAATTTGAAATGACAGC
Phenotype
MIM: 138247
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GRIA2 PubMed Links

Gene Details

Gene
GRIA2
Gene Name
glutamate ionotropic receptor AMPA type subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000826.3 Intron NP_000817.2
NM_001083619.1 Intron NP_001077088.1
NM_001083620.1 Intron NP_001077089.1
XM_017008113.1 Intron XP_016863602.1
XM_017008114.1 Intron XP_016863603.1
XM_017008115.1 Intron XP_016863604.1
XM_017008116.1 Intron XP_016863605.1
XM_017008117.1 Intron XP_016863606.1

View Full Product Details