Product Details

SNP ID
hCV44202955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:109658899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGGGGAAGAGGCCATGGTTTG[C/T]TGGAGACAAGGTAATGGGGGCATGT
Phenotype
MIM: 138380 MIM: 138333
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTM2 PubMed Links

Gene Details

Gene
GSTM2
Gene Name
glutathione S-transferase mu 2 (muscle)
There are no transcripts associated with this gene.

Gene
GSTM4
Gene Name
glutathione S-transferase mu 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000850.4 760 Missense Mutation GCT,GTT A149V NP_000841.1
NM_147148.2 760 Missense Mutation GCT,GTT A149V NP_671489.1

View Full Product Details