Product Details

SNP ID
rs11551466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47162580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTTCTGTGGGATTCTGCCCAAG[A/G]GATCTCATTACAGTTCCCAATTCCT
Phenotype
MIM: 114182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf61 PubMed Links

Gene Details

Gene
C2orf61
Gene Name
chromosome 2 open reading frame 61
There are no transcripts associated with this gene.

Gene
CALM2
Gene Name
calmodulin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305624.1 343 Silent Mutation TCC,TCT S87S NP_001292553.1
NM_001305625.1 343 Silent Mutation TCC,TCT S3S NP_001292554.1
NM_001305626.1 343 Silent Mutation TCC,TCT S3S NP_001292555.1
NM_001743.5 343 Silent Mutation TCC,TCT S39S NP_001734.1

View Full Product Details