Product Details
- SNP ID
-
rs7420190
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:25051507 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATCTCAGGAGGCCATCTGGTCAC[A/T]CTCCTTTGCGTACCATATTTTAATT
- Phenotype
-
MIM: 616797
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EFR3B
PubMed Links
Gene Details
- Gene
- EFR3B
- Gene Name
- EFR3 homolog B
View Full Product Details