Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145459.1 | 1999 | Intron | NP_001138931.1 | ||
NM_001145460.1 | 1999 | Intron | NP_001138932.1 | ||
NM_032495.5 | 1999 | Intron | NP_115884.4 | ||
NM_139211.4 | 1999 | Intron | NP_631957.1 | ||
NM_139212.3 | 1999 | Intron | NP_631958.1 | ||
XM_006714052.2 | 1999 | UTR 3 | XP_006714115.1 | ||
XM_017008728.1 | 1999 | Intron | XP_016864217.1 | ||
XM_017008729.1 | 1999 | Intron | XP_016864218.1 | ||
XM_017008730.1 | 1999 | Intron | XP_016864219.1 | ||
XM_017008731.1 | 1999 | Intron | XP_016864220.1 | ||
XM_017008732.1 | 1999 | Intron | XP_016864221.1 | ||
XM_017008733.1 | 1999 | Intron | XP_016864222.1 | ||
XM_017008734.1 | 1999 | Intron | XP_016864223.1 |