Product Details
- SNP ID
-
rs8021004
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:55030446 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGAAATGGAAGAGCCTCCTTGAC[C/T]TCAGATTTCTGAGGCACGTTTTTGT
- Phenotype
-
MIM: 616337
MIM: 608126
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SOCS4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74889539] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SOCS4
- Gene Name
- suppressor of cytokine signaling 4
- Gene
- WDHD1
- Gene Name
- WD repeat and HMG-box DNA binding protein 1
There are no transcripts associated with this gene.
View Full Product Details