Product Details

SNP ID
rs11558070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49185103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAAGGGCAGCCATGTCCTCACG[C/G]GCCTCTGAAAACTCACCTTCCTCCA
Phenotype
MIM: 602529
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TUBA1A PubMed Links

Gene Details

Gene
TUBA1A
Gene Name
tubulin alpha 1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270399.1 1696 Silent Mutation GCC,GCG A421A NP_001257328.1
NM_001270400.1 1696 Silent Mutation GCC,GCG A386A NP_001257329.1
NM_006009.3 1696 Silent Mutation GCC,GCG A421A NP_006000.2

View Full Product Details