Product Details
- SNP ID
-
rs10783409
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:50942580 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAAAATCCTTAGTTTAAAATGCAA[A/G]GCTTGGCTGGGTGTAGTGGCTCATG
- Phenotype
-
MIM: 600523
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HIGD1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111623388] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIGD1C
- Gene Name
- HIG1 hypoxia inducible domain family member 1C
- Gene
- SLC11A2
- Gene Name
- solute carrier family 11 member 2
There are no transcripts associated with this gene.
View Full Product Details