Product Details

SNP ID
rs11558752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53669260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAACTTGGCTGCTGTGTCGATGT[C/G]CCTTGAAATGGCGCTGGTTTGGAAG
Phenotype
MIM: 603193
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP5G2 PubMed Links

Gene Details

Gene
ATP5G2
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002031.2 514 Missense Mutation CAC,GAC H83D NP_001002031.1
NM_005176.5 514 Missense Mutation NP_005167.2
XM_017019460.1 514 Missense Mutation CAC,GAC H124D XP_016874949.1
XM_017019461.1 514 Missense Mutation CAC,GAC H83D XP_016874950.1
XM_017019462.1 514 Missense Mutation CAC,GAC H67D XP_016874951.1

View Full Product Details