Product Details
- SNP ID
-
rs11549158
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:116547337 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGACTCGGCTGCTTCAGTGATGA[C/T]TCCCCATGGTCAGGAATTACGGAAA
- Phenotype
-
MIM: 246600
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PNLIP
PubMed Links
Gene Details
- Gene
- PNLIP
- Gene Name
- pancreatic lipase
There are no transcripts associated with this gene.
View Full Product Details