Product Details

SNP ID
rs4987881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108223456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGGGTTCAGATTTAACTACGCG[A/G]CTTGACTAGTCATCTTTTGATCTCT
Phenotype
MIM: 607585 MIM: 601448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATM PubMed Links

Gene Details

Gene
ATM
Gene Name
ATM serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000051.3 259 Intron NP_000042.3
XM_005271561.4 259 Intron XP_005271618.2
XM_005271562.4 259 UTR 5 XP_005271619.2
XM_006718843.3 259 Intron XP_006718906.1
XM_006718845.1 259 Intron XP_006718908.1
XM_011542840.2 259 Intron XP_011541142.1
XM_011542842.2 259 Intron XP_011541144.1
XM_011542843.2 259 Intron XP_011541145.1
XM_011542844.2 259 Intron XP_011541146.1
XM_011542845.1 259 Intron XP_011541147.1
XM_017017789.1 259 Intron XP_016873278.1
XM_017017790.1 259 Intron XP_016873279.1
XM_017017791.1 259 Intron XP_016873280.1
XM_017017792.1 259 Intron XP_016873281.1
Gene
NPAT
Gene Name
nuclear protein, coactivator of histone transcription
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321307.1 259 Intron NP_001308236.1
NM_002519.2 259 Intron NP_002510.2
XM_011542854.2 259 Intron XP_011541156.1
XM_017017802.1 259 Intron XP_016873291.1
XM_017017803.1 259 Intron XP_016873292.1

View Full Product Details