Product Details

SNP ID
rs10791863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66259887 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGAGAGGAAGGCAGCATCACTC[C/T]CAAGTTTGTGATTTAAGCAAATTAA
Phenotype
MIM: 611729 MIM: 612565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLC2 PubMed Links

Gene Details

Gene
KLC2
Gene Name
kinesin light chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134774.1 Intron NP_001128246.1
NM_001134775.1 Intron NP_001128247.1
NM_001134776.1 Intron NP_001128248.1
NM_001318734.1 Intron NP_001305663.1
NM_022822.2 Intron NP_073733.1
XM_005274183.1 Intron XP_005274240.1
XM_005274184.1 Intron XP_005274241.1
XM_017018133.1 Intron XP_016873622.1
Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details