Product Details

SNP ID
rs6878299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:124643378 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTTTTGAACGCTCCATTCTTTAC[A/G]TATTAGGATAAAACAGCATCCTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF608 PubMed Links

Gene Details

Gene
ZNF608
Gene Name
zinc finger protein 608
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020747.2 Intron NP_065798.2
XM_005272037.2 Intron XP_005272094.1
XM_005272038.1 Intron XP_005272095.1
XM_005272039.2 Intron XP_005272096.1
XM_005272040.3 Intron XP_005272097.1
XM_011543520.2 Intron XP_011541822.1
XM_011543522.2 Intron XP_011541824.1
XM_011543523.2 Intron XP_011541825.1
XM_017009674.1 Intron XP_016865163.1

View Full Product Details