Product Details

SNP ID
rs11945475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:58837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTCTGCTTTCTCTAGCTGAGT[A/G]GTACTATAACGGGTCTAGGAGGAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF595 PubMed Links

Gene Details

Gene
ZNF595
Gene Name
zinc finger protein 595
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286052.1 Intron NP_001272981.1
NM_001286053.1 Intron NP_001272982.1
NM_001286054.1 Intron NP_001272983.1
NM_182524.3 Intron NP_872330.1
XM_011513398.2 Intron XP_011511700.1

View Full Product Details