Product Details
- SNP ID
-
rs8033750
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:20670172 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATATATATATAATCTGCAATTAA[C/T]GTTTTCTAATAAACTTATTATAATA
- Phenotype
-
MIM: 601889
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NBEAP1
PubMed Links
Gene Details
- Gene
- NBEAP1
- Gene Name
- neurobeachin pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details