Product Details

SNP ID
hCV45179965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70033394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGTTAGCTCTTTGAAGTGTGTT[A/G]ATTTATATAGATGCTTCTGACTTTG
Phenotype
MIM: 142702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTN3 PubMed Links

Gene Details

Gene
HTN3
Gene Name
histatin 3
There are no transcripts associated with this gene.

View Full Product Details