Product Details
- SNP ID
-
rs694031
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:30606533 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATCCCCAGCTGAATTAGTCACCAA[C/T]GCTTTTTAAAAGCCTGGTTTGTGCC
- Phenotype
-
MIM: 612748
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LYZL2
PubMed Links
Gene Details
- Gene
- LYZL2
- Gene Name
- lysozyme like 2
View Full Product Details