Product Details
- SNP ID
-
rs6604980
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
36
- Location
-
Chr.1:1659065 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTCCGGCCCAGAACTCGAGACAA[C/G]GACAGGGGCTCGCTCTGTGCGGCAC
- Phenotype
-
MIM: 176873
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDK11B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79113395] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDK11B
- Gene Name
- cyclin dependent kinase 11B
- Gene
- SLC35E2B
- Gene Name
- solute carrier family 35 member E2B
View Full Product Details