Product Details

SNP ID
rs4970459
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:922671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGCTTCTGGGCATGTTCCTTG[C/T]CTGAGCAGAGCACTCCTGGCCCAGA
Phenotype
MIM: 616765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130417 PubMed Links

Gene Details

Gene
LOC100130417
Gene Name
uncharacterized LOC100130417
There are no transcripts associated with this gene.

Gene
LOC107985728
Gene Name
collagen alpha-1(III) chain-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017003065.1 1988 UTR 3 XP_016858554.1
Gene
LOC284600
Gene Name
uncharacterized LOC284600
There are no transcripts associated with this gene.

Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
There are no transcripts associated with this gene.

View Full Product Details