Product Details

SNP ID
rs3121571
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:1000112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGCCCGCGCGCGCCTCAGGCC[G/T]TTTCCCTATTTAAGGCCTCGCCGCC
Phenotype
MIM: 608060
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HES4 PubMed Links

Gene Details

Gene
HES4
Gene Name
hes family bHLH transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142467.1 941 UTR 5 NP_001135939.1
NM_021170.3 941 UTR 5 NP_066993.1
XM_005244771.4 941 UTR 5 XP_005244828.1

View Full Product Details