Product Details

SNP ID
rs1127513
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:151364693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTCTACATCAACCTGCAGCAT[C/T]ACAGAGCCTTCCCTGGTGGAAAAGG
Phenotype
MIM: 604188
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SELENBP1 PubMed Links

Gene Details

Gene
SELENBP1
Gene Name
selenium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258288.1 1571 Silent Mutation GTA,GTG V361V NP_001245217.1
NM_001258289.1 1571 Silent Mutation GTA,GTG V465V NP_001245218.1
NM_003944.3 1571 Silent Mutation GTA,GTG V423V NP_003935.2
XM_017002757.1 1571 Intron XP_016858246.1

View Full Product Details