Product Details
- SNP ID
-
rs28438930
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:171842981 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGCTGGCGAGATTGTACTTTAT[C/T]CTTTTTATTTGGCTCATTCGTGACT
- Phenotype
-
MIM: 611445
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNM3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114532899] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNM3
- Gene Name
- dynamin 3
View Full Product Details