Product Details

SNP ID
rs28362635
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:155688648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCAAGCCGGCTCCAGCGCAGGCC[C/T]TCACGCGTACCTTCAGCGGCGCGAG
Phenotype
MIM: 602074 MIM: 607860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAP3 PubMed Links

Gene Details

Gene
DAP3
Gene Name
death associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199849.1 9 Intron NP_001186778.1
NM_001199850.1 9 Intron NP_001186779.1
NM_001199851.1 9 Intron NP_001186780.1
NM_004632.3 9 Intron NP_004623.1
NM_033657.2 9 Intron NP_387506.1
XM_005245480.2 9 Intron XP_005245537.1
XM_005245481.2 9 Intron XP_005245538.1
XM_005245482.4 9 Intron XP_005245539.1
XM_011509972.2 9 Intron XP_011508274.1
XM_017002289.1 9 Intron XP_016857778.1
XM_017002290.1 9 Intron XP_016857779.1
XM_017002291.1 9 Intron XP_016857780.1
XM_017002292.1 9 Intron XP_016857781.1
XM_017002293.1 9 Intron XP_016857782.1
XM_017002294.1 9 Intron XP_016857783.1
XM_017002295.1 9 Intron XP_016857784.1
XM_017002296.1 9 Intron XP_016857785.1
XM_017002297.1 9 Intron XP_016857786.1
Gene
SCARNA26A
Gene Name
small Cajal body-specific RNA 26A
There are no transcripts associated with this gene.

Gene
YY1AP1
Gene Name
YY1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198899.1 9 Intron NP_001185828.1
NM_001198900.1 9 Intron NP_001185829.1
NM_001198901.1 9 Intron NP_001185830.1
NM_001198902.1 9 Intron NP_001185831.1
NM_001198903.1 9 UTR 5 NP_001185832.1
NM_001198904.1 9 UTR 5 NP_001185833.1
NM_001198905.1 9 Intron NP_001185834.1
NM_001198906.1 9 Intron NP_001185835.1
NM_018253.3 9 Intron NP_060723.2
NM_139118.2 9 Intron NP_620829.1
NM_139119.2 9 Intron NP_620830.1
NM_139121.2 9 Intron NP_620832.1

View Full Product Details