Product Details

SNP ID
rs3093566
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:25338442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACACCGGGCTCCGGAACCACTGC[A/T]CGACGGGGCTGGACGTAAGTGTCGG
Phenotype
MIM: 111680 MIM: 605348
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RHD PubMed Links

Gene Details

Gene
RHD
Gene Name
Rh blood group D antigen
There are no transcripts associated with this gene.

Gene
RSRP1
Gene Name
arginine and serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321772.1 145 Intron NP_001308701.1
NM_020317.4 145 Intron NP_064713.3
XM_011541797.1 145 Intron XP_011540099.1
Gene
TMEM50A
Gene Name
transmembrane protein 50A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014313.3 145 UTR 5 NP_055128.1
XM_005245817.1 145 UTR 5 XP_005245874.1
XM_011541159.1 145 Intron XP_011539461.1

View Full Product Details