Product Details

SNP ID
rs6587483
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:248500920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGATAAGAAATGCAGGTAATCA[A/G]AACTCTCCTTCCTGCCCTCTTCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2T10 PubMed Links

Gene Details

Gene
OR2T10
Gene Name
olfactory receptor family 2 subfamily T member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004693.1 Intron NP_001004693.1
XM_017000273.1 Intron XP_016855762.1

View Full Product Details