Product Details
- SNP ID
-
rs28395444
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:239388185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGCAAACCTTCATTTGTCTGCAA[A/G]TGTTACTGGAACCTGTCCAAAGGCT
- Phenotype
-
MIM: 118494
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHRM3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76484712] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHRM3
- Gene Name
- cholinergic receptor muscarinic 3
- Gene
- LOC107983951
- Gene Name
- uncharacterized LOC107983951
View Full Product Details