Product Details

SNP ID
rs28796397
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:16380992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCAGCTGCTCAGGAGGCTGAGG[C/G]AGATCGCTTGAGTCCAGGAAATCAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SZRD1 PubMed Links

Gene Details

Gene
SZRD1
Gene Name
SUZ RNA binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114600.2 Intron NP_001108072.1
NM_001271869.1 Intron NP_001258798.1

View Full Product Details