Product Details

SNP ID
rs11547938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:182717056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAAGGAAGTATTAGAAATGAG[C/G]TGAAGACCATTCACAGGTAAAGAGA
Phenotype
MIM: 607987
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC10 PubMed Links

Gene Details

Gene
DNAJC10
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271581.1 484 UTR 5 NP_001258510.1
NM_018981.2 484 UTR 5 NP_061854.1

View Full Product Details