Product Details

SNP ID
rs28539487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197468089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCAAAACATAACAGTAAAAATCC[A/G]TGCGTCCTTGGGCTAGATAATTGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ10B PubMed Links

Gene Details

Gene
COQ10B
Gene Name
coenzyme Q10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320818.1 Intron NP_001307747.1
NM_001320819.1 Intron NP_001307748.1
NM_001320820.1 Intron NP_001307749.1
NM_025147.4 Intron NP_079423.1

View Full Product Details