Product Details

SNP ID
rs35910919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218959696 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGGAGCCGCCTAGCAGCCACCTT[A/C]CCCCGCCAGTCCGCGCGCCCGGGCC
Phenotype
MIM: 603764
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDK5R2 PubMed Links

Gene Details

Gene
CDK5R2
Gene Name
cyclin dependent kinase 5 regulatory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003936.4 69 UTR 5 NP_003927.1

View Full Product Details