Product Details

SNP ID
rs35742624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223977317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCTCTGTTAAAAACAACAATC[C/T]GCAAAACCTAAAATCCTGTCAGTGT
Phenotype
MIM: 611849 MIM: 177010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL44 PubMed Links

Gene Details

Gene
MRPL44
Gene Name
mitochondrial ribosomal protein L44
There are no transcripts associated with this gene.

Gene
SERPINE2
Gene Name
serpin family E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136528.1 Intron NP_001130000.1
NM_001136530.1 Intron NP_001130002.1
NM_006216.3 Intron NP_006207.1
XM_005246641.2 Intron XP_005246698.1
XM_017004329.1 Intron XP_016859818.1
XM_017004330.1 Intron XP_016859819.1
XM_017004331.1 Intron XP_016859820.1
XM_017004332.1 Intron XP_016859821.1

View Full Product Details