Product Details
- SNP ID
-
rs28660615
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:239898899 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCTCCACATGTAAAGGGGTGTGA[A/T]GGAGGGGAGTGATGGAAGGTTGTGA
- Phenotype
-
MIM: 603835
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NDUFA10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75499001] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NDUFA10
- Gene Name
- NADH:ubiquinone oxidoreductase subunit A10
View Full Product Details