Product Details

SNP ID
rs34850174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241496172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCTGAGACACTGCCTCAGACC[C/G]TTAGACTGGAAAGGCCCTGCCCACT
Phenotype
MIM: 602255
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FARP2 PubMed Links

Gene Details

Gene
FARP2
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 2
There are no transcripts associated with this gene.

Gene
STK25
Gene Name
serine/threonine kinase 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271977.1 Intron NP_001258906.1
NM_001271978.1 Intron NP_001258907.1
NM_001271979.1 Intron NP_001258908.1
NM_001271980.1 Intron NP_001258909.1
NM_001282305.1 Intron NP_001269234.1
NM_001282306.1 Intron NP_001269235.1
NM_001282307.1 Intron NP_001269236.1
NM_001282308.1 Intron NP_001269237.1
NM_006374.4 Intron NP_006365.2
XM_011510493.2 Intron XP_011508795.1
XM_011510494.2 Intron XP_011508796.1
XM_011510495.2 Intron XP_011508797.1
XM_011510496.2 Intron XP_011508798.1
XM_017003168.1 Intron XP_016858657.1
XM_017003169.1 Intron XP_016858658.1
XM_017003170.1 Intron XP_016858659.1
XM_017003171.1 Intron XP_016858660.1

View Full Product Details