Product Details

SNP ID
rs13016102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241870972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCCACTGCCTGGAACTGAATCA[C/T]AGCCTCTCACTCATGCAGGGTTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTP5 PubMed Links

Gene Details

Gene
RTP5
Gene Name
receptor transporter protein 5 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173821.2 993 Intron NP_776182.2
XM_011510995.2 993 UTR 5 XP_011509297.1
XM_011510997.2 993 Intron XP_011509299.1

View Full Product Details