Product Details

SNP ID
rs7420371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241871857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCAGGCTGTGCCCCGCACCCG[A/G]GGACTGCCAGGTGAGGCCCCCGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01237 PubMed Links

Gene Details

Gene
LINC01237
Gene Name
long intergenic non-protein coding RNA 1237
There are no transcripts associated with this gene.

Gene
RTP5
Gene Name
receptor transporter protein 5 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173821.2 325 Missense Mutation GAG,GGG E101G NP_776182.2
XM_011510995.2 325 Missense Mutation GAG,GGG E80G XP_011509297.1
XM_011510997.2 325 Missense Mutation GAG,GGG E101G XP_011509299.1

View Full Product Details