Product Details
- SNP ID
-
rs28521658
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:15973949 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGATGAGGCAGGGTGGTCATCCT[A/G]CCCGGTTCCTGGGGCTGCTGGACCA
- Phenotype
-
MIM: 604365
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROM1
PubMed Links
Gene Details
- Gene
- PROM1
- Gene Name
- prominin 1
View Full Product Details