Product Details

SNP ID
rs28383118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:83078025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGATATTGGTAAACTGTGGGTT[C/G]CAACCAGGCATCGAGGCTGAAACGT
Phenotype
MIM: 194363
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM167A PubMed Links

Gene Details

Gene
TMEM167A
Gene Name
transmembrane protein 167A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174909.4 Intron NP_777569.1
Gene
XRCC4
Gene Name
X-ray repair cross complementing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318012.1 Intron NP_001304941.1
NM_001318013.1 Intron NP_001304942.1
NM_003401.4 Intron NP_003392.1
NM_022406.3 Intron NP_071801.1
NM_022550.3 Intron NP_072044.1
XM_011543626.1 Intron XP_011541928.1
XM_017009827.1 Intron XP_016865316.1
XM_017009828.1 Intron XP_016865317.1
XM_017009829.1 Intron XP_016865318.1

View Full Product Details