Product Details
- SNP ID
-
hCV58519998
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:87086413 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCTGGGAGAAGAATGGGTTTTCC[C/T]GTAGCGTGCATTCTGGGCAATCTAT
- Phenotype
-
MIM: 118850
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CGA
PubMed Links
Gene Details
- Gene
- CGA
- Gene Name
- glycoprotein hormones, alpha polypeptide
There are no transcripts associated with this gene.
View Full Product Details