Product Details

SNP ID
rs28381466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87594261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCTCCCCTAACATCTTTTTTT[A/T]AAAAAGTGCTTACCCATAAAATATT
Phenotype
MIM: 604972 MIM: 611524
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ORC3 PubMed Links

Gene Details

Gene
ORC3
Gene Name
origin recognition complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197259.1 Intron NP_001184188.1
NM_012381.3 Intron NP_036513.2
NM_181837.2 Intron NP_862820.1
XM_005248704.1 Intron XP_005248761.1
XM_005248705.1 Intron XP_005248762.1
XM_005248706.1 Intron XP_005248763.1
XM_011535651.1 Intron XP_011533953.1
XM_011535652.2 Intron XP_011533954.1
XM_017010632.1 Intron XP_016866121.1
XM_017010633.1 Intron XP_016866122.1
XM_017010634.1 Intron XP_016866123.1
XM_017010635.1 Intron XP_016866124.1
XM_017010636.1 Intron XP_016866125.1
XM_017010637.1 Intron XP_016866126.1
XM_017010638.1 Intron XP_016866127.1
XM_017010639.1 Intron XP_016866128.1
XM_017010640.1 Intron XP_016866129.1
Gene
RARS2
Gene Name
arginyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details